Falcon™ Tissue Culture-Treated 96-well Microplates
Polystyrene microplates with tissue culture-treated surface
$555.74 - $555.74
Specifications
For Use With (Application) It supports a wide range of applications and many important cell types, including primary, stem, neuronal, mesenchymal, hepatocyte and endothelial cells
Material Polystyrene
Sterile Yes
Type 96-Well Assay Microplate
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
BD353377 Corning Life Sciences
C353377
White Case of 32 Each for $555.74
Specifications
Description
  • Flat bottom; with lid
  • Storage: -20° to +60°C
  • Sterile; Class I Medical
  • Choose black or white/clear
  • Surface area: 10 sq. cm
  • 8 per pack; 32 per case
Description & Specifications
Specifications
It supports a wide range of applications and many important cell types, including primary, stem, neuronal, mesenchymal, hepatocyte and endothelial cells
Polystyrene
Yes
96-Well Assay Microplate