Please login to your online account to display your discounted pricing

Falcon™ Polystyrene Microplates


Constructed of crystal grade virgin polystyrene for optimum visibility. Corning™ Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.

Manufacturer: corning life sciences  C353077

 View more versions of this product

Catalog No. 08-772-17

  • / Case of 50 Each
 

Description & Specifications

Specifications

Lid Yes
Surface Coating Tissue-culture treated
Type Cell Culture Microplate
Volume (Metric) Working 1.5 to 2.2mL
Color Clear
Culture Area 0.36cm2
Format 96-well
Material Polystyrene
No. of Wells 96
Quantity 1/Tray, 50/Cs.
Sterile Yes
Volume (Metric) 0.32mL
Well Design Round
Closure Yes
Material Polystyrene
Format 96-well
Volume (Metric) 0.32mL
Type Cell Culture Microplate
Lid Yes
Wells 96
Well Shape Round

  • Choose tissue culture-treated or nontreated surfaces
  • Nontreated growth surface is more hydrophobic than tissue culture surface and is similiar to the surface of a bacteriological Petri dish
  • Sterilized by gamma irradiation
  • Lid design permits stable stacking on benchtop or in incubator
  • All plates (except 08-772-2A and 08-772-16) have patented “labyrinth” lid that reduces evaporation of media from wells, minimizes the risk of contamination
  • Deep skirt and serrated edge permit a firmer grip for plate manipulation
  • Exceptional optical clarity allows for visualization of all cell monolayers
  • Frosted writing ends provide clear writing area for quick identification
  • All plates and lids are suitable for both manual and automated handling