missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Polystyrene Microplates

Constructed of crystal grade virgin polystyrene for optimum visibility. Corning™ Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.

Manufacturer:  Corning Life Sciences C353227

 View more versions of this product

Catalog No. 08-772-3B


 

Specifications

Description

Microplate, Cell Culture; Corning; Falcon; 96-well; U-bottom with lid; Standard tissue culture; Clear polystyrene; Sterile; Growth area: 0.36cm2; Well volume: 0.32mL; Working volume: 1.5-2.2mL; 5/Bag

  • Choose tissue culture-treated or nontreated surfaces
  • Nontreated growth surface is more hydrophobic than tissue culture surface and is similiar to the surface of a bacteriological Petri dish
  • Sterilized by gamma irradiation
  • Lid design permits stable stacking on benchtop or in incubator
  • All plates (except 08-772-2A and 08-772-16) have patented “labyrinth” lid that reduces evaporation of media from wells, minimizes the risk of contamination
  • Deep skirt and serrated edge permit a firmer grip for plate manipulation
  • Exceptional optical clarity allows for visualization of all cell monolayers
  • Frosted writing ends provide clear writing area for quick identification
  • All plates and lids are suitable for both manual and automated handling

Description & Specifications

Specifications

U-bottom
Clear
96-well
96
Yes
Cell Culture Microplate
1.5 to 2.2mL
Yes
0.36cm2
Polystyrene
5/Bag, 50/Cs.
Tissue-culture treated
0.32mL
Yes