missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Sterile Sample Containers

Made of clear polypropylene for strength and visibility. Corning Falcon™ Sterile Sample Containers are ideal for specimen collection.

Manufacturer:  Falcon™ 354015

 View more versions of this product

Catalog No. 11-838-7



Description

Description

  • Sterile, disposable polypropylene containers provide secure sample containment
  • Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
  • Feature molded-in graduations for easy measurements
  • Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
  • Features

    Sterile
  • 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
  • 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
Specifications

Specifications

220 mL
Yes
Sterile
100/Cs.
Yes
Polypropylene
1/Pk.
SDS
Documents
Product Certifications
Promotions

Promotions