Please login to your online account to display your discounted pricing

Falcon™ Standard Tissue Culture Dishes


Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.

Manufacturer: corning life sciences  C353002

 View more versions of this product

Catalog No. 08-772B

  • / Case of 500 Each
 

Description & Specifications

Specifications

Material Polystyrene
Bottom Shape Round (U)
Surface Area (Metric) 21.29cm2
Height (Metric) Exterior 15mm
Diameter (Metric) 60mm
Sterile Yes
Surface Coating TC
Volume (Metric) Working 6 to 7mL

  • Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
  • Improved handling of small dishes with the unique Easy-Grip design
  • Nonpyrogenic
  • Sterilized by gamma irradiation
  • Packaged in peel-open, medical-style bags
  • Crystal-grade virgin polystyrene