missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Serological Pipets

Features exceptionally crisp, dark, easy-to-read printing with reverse and negative graduations. Corning™ Falcon™ Serological Pipets are individually wrapped in Advantage™ all-plastic thermoformed package.

Manufacturer:  Corning Life Sciences C356535

 View more versions of this product

Catalog No. 13-675-51



Description

Description

 

  • Gamma-irradiated, nonpyrogenic, made of clear polystyrene for optical clarity
  • Exclusive graduation alignment mark allows graduations to be brought into line of sight before complete removal of the wrapper
  • Inside surface discourages formation of meniscus, thereby reducing reading errors Total delivery accuracy of pipets, ±3%, with a 1.5% coefficient of variation
  • Negative graduations allow for extra capacity
  • Polyester fiber plug helps prevent overfill
  • Color coding on packaging and blow-out rings allows easy identification of size

 

Specifications

Specifications

0.84 oz.
1.5%
Yes
0.5mL
Individually wrapped
Falcon™ Serological Pipet
PS (Polystyrene)
0.84 oz., 25mL
Purple
Yes
11mL
Standard
25mL
Documents
Accessories