Falcon™ Serological Pipets
Features exceptionally crisp, dark, easy-to-read printing with reverse and negative graduations. Corning™ Falcon™ Serological Pipets are individually wrapped in Advantage™ all-plastic thermoformed package.
Manufacturer:  Corning Life Sciences C356525
 View more versions of this product
Catalog No. 13-675-50
Description
Description
  • Gamma-irradiated, nonpyrogenic, made of clear polystyrene for optical clarity
  • Exclusive graduation alignment mark allows graduations to be brought into line of sight before complete removal of the wrapper
  • Inside surface discourages formation of meniscus, thereby reducing reading errors Total delivery accuracy of pipets, ±3%, with a 1.5% coefficient of variation
  • Negative graduations allow for extra capacity
  • Polyester fiber plug helps prevent overfill
  • Color coding on packaging and blow-out rings allows easy identification of size
Specifications
Specifications
±3%
0.84 oz.
1.5%
Yes
0.25mL
Individually wrapped
Falcon™ Serological Pipet
PS (Polystyrene)
Yes
0.84 oz., 25mL
Purple
Yes
7mL
Standard
25mL
Documents
Accessories