Please login to your online account to display your discounted pricing
Falcon™ Pipet Controller Hydrophobic PTFE Filters
Replacement hydrophobic PTFE disc filters
Manufacturer: corning life sciences  C357472
 View more versions of this product
Catalog No. 05-100-502
  • / Case of 5 Each
Description & Specifications
Specifications
For Use With (Equipment) For use with pipet controller
Material PTFE
Pore Size 0.2μm
Quantity 5/Cs.
For Use With (Equipment) For use with pipet controller