Please login to your online account to display your discounted pricing

Falcon™ Multiwell Insert System


Ideal for both manual and robotic screening of compounds in cell-based assays

Manufacturer: corning incorporated  C351130

 View more versions of this product

Catalog No. 08-771-000

  • / Case
 

Description & Specifications

Specifications

Lid Yes
Culture Area 0.08cm2
Format 96-well
For Use With (Application) To produce a differentiated monolayer of Caco-2 cells, suitable for bioavailability and permeability studies
Pore Size 1μm
Sterile Yes
No. of Wells 96
Membrane Polyethylene Terephthalate
Closure Yes
Color Translucent
Type Multiwell Insert System
Includes One insert plate with feeder tray and lid