Falcon™ Multiwell Insert System
Ideal for both manual and robotic screening of compounds in cell-based assays
$913.92 - $4,142.00
Specifications
Culture Area 0.08cm2
Color Translucent
Sterile Yes
Format 96-well
Pore Size 1μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
08-771-000 Corning Incorporated
C351130
One insert plate with feeder tray and lid Case for $913.92
08-771-001 Corning Incorporated
C351131
Five insert plates with feeder trays and lids Case of 5 Each for $4,142.00
Specifications
Description
One-piece, 96-well polyethylene terephthalate (PET) insert plate with 1μm pore size PET membrane fits into feeder tray, which distributes media uniformly to each well. Individual sample conditions can be tested using the 96–Square-Well Angled-Bottom Plate (sold separately).
•    96 angled-bottom wells permit complete aspiration or collection of samples from the bottom of the well
•    Insert plate has generous, automation-compatible sampling ports positioned directly above the angled well bottom to facilitate sampling above and below the membrane
•    Tested for the ability to produce a differentiated monolayer of Caco-2 cells. Suitable for bioavailability and permeability studies
•    Insert Systems include insert, feeder tray, and lid; additional feeder trays can be purchased separately
•    All products are gamma-irradiation sterilized
Note: Standard Falcon 96-well plates are not compatible with the HTS 96-Multiwell Insert System; use of standard 96-well plates will result in media wicking on the sides of the well and possibly into the insert or out of the well
Description & Specifications
Specifications
0.08cm2
Yes
1μm
96
Translucent
96-well
Multiwell Insert System