Please login to your online account to display your discounted pricing

Falcon™ Erlenmeyer Flasks


High performance polycarbonate Erlenmeyer flask for suspension culture and storage

Manufacturer: corning life sciences  C355115

 View more versions of this product

Catalog No. 08-772-250

  • / Case of 24 Each
 

Description & Specifications

Specifications

Capacity (English) 4.22 oz.
Capacity 4.22 oz., 125mL
Bottom Shape Baffled
Capacity (Metric) 125mL
Disposable Yes
For Use With (Application) Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
Material Polycarbonate
Storage Requirements 15 to 30°C
Sterile Yes