Please login to your online account to display your discounted pricing

Falcon™ Erlenmeyer Flasks


High performance polycarbonate Erlenmeyer flask for suspension culture and storage

Manufacturer: corning life sciences  C355129

 View more versions of this product

Catalog No. 08-772-258

  • / Case of 6 Each
 

Description & Specifications

Specifications

Storage Requirements 15 to 30°C
Capacity 33.8 oz., 1000mL
Disposable Yes
Capacity (Metric) 1000mL
Material Polycarbonate
Capacity (English) 33.8 oz.
Bottom Shape Flat
For Use With (Application) Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
Sterile Yes