Falcon™ Erlenmeyer Flasks
High performance polycarbonate Erlenmeyer flask for suspension culture and storage
$437.59 - $760.23
Specifications
Material Polycarbonate
Sterility Sterile
Disposable Yes
For Use With (Application) Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
Description Store at: 15 to 30°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Bottom Shape Price Quantity  
Catalog Number Mfr. No. Bottom Shape Price Quantity  
08-772-250 Corning Life Sciences
C355115
Baffled Case of 24 Each for $760.23
08-772-251 Corning Life Sciences
C355119
Baffled Case of 12 Each for $440.31
08-772-253 Corning Life Sciences
C355127
Baffled Case of 6 Each for $437.59
08-772-257 Corning Life Sciences
C355125
Flat Case of 12 Each for $646.28
08-772-258 Corning Life Sciences
C355129
Flat Case of 6 Each for $448.18
Specifications
Description
Falcon Erlenmeyer flasks are disposable culture and storage vessels for cell biology, microbiology, or molecular biology applications. They are ideal for use on shaker equipment for culturing bacterial, yeast, and mammalian cells as well as for reagent storage.
  • Provide flexibility with a 2-in-1 vented or non-vented leakproof cap
  • Allow uninhibited visualization of sample or media through the optically clear polycarbonate resin, which is lighter and safer than glass
  • Non-pyrogenic, non-cytotoxic, and nuclease-free
  • The wide mouth design of these flasks allows quick and easy fluid additions and removal, as well as providing easy pipet access to the bottom edges
  • The molded-in graduations provide accurate volumetric measurement for media and buffers
  • 0.2μm PTFE membrane allows for sterile air transfer during aerobic applications
  • Ready-to-use, individually packaged, autoclavable, sterile to SAL 10-6
  • Available in flat- or baffled-bottom, which provides increased agitation and aeration
Compliance
USP Class VI
Description & Specifications
Specifications
Polycarbonate
Yes
Store at: 15 to 30°C
Sterile
Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor