Please login to your online account to display your discounted pricing

Falcon™ Erlenmeyer Flasks


High performance polycarbonate Erlenmeyer flask for suspension culture and storage

Manufacturer: corning life sciences  C355119

 View more versions of this product

Catalog No. 08-772-251

  • / Case of 12 Each
 

Description & Specifications

Specifications

Capacity (English) 8.45 oz.
Capacity (Metric) 250mL
Material Polycarbonate
Sterile Yes
Capacity 8.45 oz., 250mL
Bottom Shape Baffled
Disposable Yes
For Use With (Application) Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
Storage Requirements 15 to 30°C