Corning™ Falcon™ Easy-Grip Tissue Culture Dishes
Features flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning™ Falcon™ Easy-Grip Tissue Culture Dishes are uniquely designed to simplify lifting one dish or a small stack of dishes.
Manufacturer:  Corning™ C353004
 View more versions of this product
Catalog No. 08-772F
Specifications
Description
Dish, Cell Culture; Corning; Falcon; Standard TC; 60 x 15mm; Easy-Grip; Growth area: 19.5cm2; Working volume: 6.0 to 7.0mL; 20/pk, 500/cs
  • Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
  • Improved handling of small dishes with the unique Easy-Grip design
  • Nonpyrogenic
  • Sterilized by gamma irradiation
  • Packaged in peel-open, medical-style bags
  • Crystal-grade virgin polystyrene
Description & Specifications
Specifications
60mm
Yes
19.5cm2
6 to 7mL
Polystyrene