Please login to your online account to display your discounted pricing

Falcon™ Cell Strainers


Rapidly isolates primary cells to consistently obtain a uniform single-cell suspension from tissues. Corning Falcon™ Cell Strainers are easy to use and ideal for use with stem cells and primary cells.

Manufacturer: corning life sciences  C352360

 View more versions of this product

Catalog No. 08-771-19

  • / Case of 50 Each
 

Description & Specifications

Specifications

Product Type Cell Strainer
Disposable Yes
Mesh Size 100μm
For Use With (Equipment) Falcon 50mL conical tubes
Color Yellow
Material Polypropylene
For Use With (Application) Designed for rapidly isolating primary cells to consistently obtain a uniform single cell suspension from tissues
Mesh Material Nylon
Includes Extended lip on strainer, polypropylene frame with tab
Sterile Yes

 

Features
  • Strong nylon mesh with 40, 70, and 100μm micron pores for optimal performance in a variety of applications
  • Evenly spaced mesh pores providing consistent and reliable results
  • Conveniently accessible in individual packaging
  • Extended lip on strainer enables aseptic handling with forceps
  • Molded color-coded polypropylene frame with tab enables easy handling and identification
  • Fits perfectly into 50mL Falcon conical tubes
  • Disposable, easy-to-use, inexpensive, maintains sample integrity
  • Sterilized by gamma irradiation, noncytotoxic
Application
  • Faster, easier alternative to gauze filtration
  • Ideal for preparation of flow cytometry samples, including: 
    • Single cell suspensions of blood cells from marrow, pancreas, thymus, tonsil, and lymph nodes
    • Stem cells, tissue-derived cells, and cancer cells 
  • Preparation of specimens for primary cell cultures and immunogens  
  • Preparation of freezing stocks 
  • Filtering agglutinative proteins produced in inactivation serum