missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Cell Strainers

Rapidly isolates primary cells to consistently obtain a uniform single-cell suspension from tissues. Corning Falcon™ Cell Strainers are easy to use and ideal for use with stem cells and primary cells.

Manufacturer:  Corning Life Sciences C352340

 View more versions of this product

Catalog No. 08-771-1


 

Specifications

Description

Strainer, Cell; Corning; Falcon; Nylon mesh; For use with Falcon 50mL conical tubes; Individually wrapped; Sterile; Pore size: 40um; Color: blue

 

Features
  • Strong nylon mesh with 40, 70, and 100μm micron pores for optimal performance in a variety of applications
  • Evenly spaced mesh pores providing consistent and reliable results
  • Conveniently accessible in individual packaging
  • Extended lip on strainer enables aseptic handling with forceps
  • Molded color-coded polypropylene frame with tab enables easy handling and identification
  • Fits perfectly into 50mL Falcon conical tubes
  • Disposable, easy-to-use, inexpensive, maintains sample integrity
  • Sterilized by gamma irradiation, noncytotoxic
Application
  • Faster, easier alternative to gauze filtration
  • Ideal for preparation of flow cytometry samples, including: 
    • Single cell suspensions of blood cells from marrow, pancreas, thymus, tonsil, and lymph nodes
    • Stem cells, tissue-derived cells, and cancer cells 
  • Preparation of specimens for primary cell cultures and immunogens  
  • Preparation of freezing stocks 
  • Filtering agglutinative proteins produced in inactivation serum
Description & Specifications

Specifications

Yes
Falcon 50mL conical tubes
Polypropylene
40μm
Blue
Designed for rapidly isolating primary cells to consistently obtain a uniform single cell suspension from tissues
Extended lip on strainer, polypropylene frame with tab
Nylon
Cell Strainer
Yes