missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Cell Culture Inserts: For Use With 6-Well Plates

Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types

Manufacturer:  Corning Life Sciences C353092

 View more versions of this product

Catalog No. 08-771-6


 

Specifications Description & Specifications

Specifications

4.2cm2
Yes
3μm
6 Well Plate
PET
2.0 x 106 (HD)
Translucent Membrane
23.1mm