Please login to your online account to display your discounted pricing

Falcon™ Cell Culture Inserts: For Use With 6-Well Plates


Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types

Manufacturer: corning life sciences  C353093

 View more versions of this product

Catalog No. 08-771-20

  • / Case of 48 Each
 

Description & Specifications

Specifications

Diameter (Metric) Filter 23.1mm
Type Translucent Membrane
Material PET
Culture Area 4.2cm2
For Use With (Equipment) 6 Well Plate
Pore Size 8μm
Sterile Yes