Please login to your online account to display your discounted pricing
Falcon™ Cell Culture Inserts: For Use With 12-Well Plates
Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types
Manufacturer: corning life sciences  C353180
 View more versions of this product
Catalog No. 08-771-10
  • / Case of 48 Each
Description & Specifications
Specifications
Culture Area 0.9cm2
Material PET
Sterile Yes
Pore Density 1.6 x 106
Pore Size 0.4μm
Type Transparent Membranes
For Use With (Equipment) 12 Well Plate
Diameter (Metric) Filter 10.5mm