missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Cell Culture Inserts: For Use With 12-Well Plates

Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types

Manufacturer:  Corning Life Sciences C353103

 View more versions of this product

Catalog No. 08-771-8


 

Specifications Description & Specifications

Specifications

0.9cm2
Yes
1μm
12 Well Plate
PET
1.6 x 106
Transparent Membranes
10.5mm