Falcon™ 96-Well Assay Plates
Manufactured from crystal-grade virgin polystyrene for distortion-free optics. Corning™ Falcon™ 96-Well Assay Plates are injection-molded and compatible with automation systems.
$327.26 - $580.77
Specifications
Color Clear
Sterile No
Type Cell Culture Microplate
No. of Wells 96
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Volume (Metric) Well Price Quantity  
Catalog Number Mfr. No. Material Volume (Metric) Well Price Quantity  
08-772-212 Corning Life Sciences
C353263
Polypropylene 250μL Case of 100 Each for $580.77
08-772-5 Corning Life Sciences
C353910
Polystyrene 320μL Case of 50 Each for $327.26
Specifications
Description
  • Standard untreated polystyrene plates are hydrophobic
  • Easy-grip serrated base (on long sides only) compatible with human and robotic hands
  • Alphanumeric notation system for well identification
  • Meet standard ANSI/SBS footprint dimensions for microplates
  • Nonpyrogenic
  • Convenient, peel-open medical-style packaging for individual product presentation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Description & Specifications
Specifications
Clear
No
Cell Culture Microplate
96