missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ FlouroBlok™96 Well HTS Insert Systems

Simplify detection of migrating fluorescently labeled cells. Corning™ Falcon™ FlouroBlok™96 Well HTS Insert Systems are designed with a unique light-tight PET membrane that blocks the transmission light within the range of 400 to 700nm.

Manufacturer:  Corning Incorporated C351163

 View more versions of this product

Catalog No. 08-771-006


 

Specifications

Description

Microplates, 96-Multiwell insert system; BD Falcon FluoroBlok; With 96-well black plate and lid; 8μm pore size; Compatible with standard fluorescence plate readers, robots, and fluid handling systems; 1/cs

 

  • Homogeneous fluorescence assay system
  • Collect data without plate washing, cell scraping, and counting
  • Chart cell migration or bioavailability in real time without dismantling or destroying the insert
  • Each insert is fitted with a fluorescence blocking Corning Fluoroblok membrane
  • Migrated cells below the membrane are easily detected by a fluorescence plate reader while those remaining above the membrane are made “invisible”
  • Migrated cells are no longer shielded from the light source and are easily detected with a fluorescence plate reader or inverted fluorescence microscope
  • Store at room temperature, away from direct sunlight.

Applications
  • Study a wide range of cell types and activities such as: 
  • Inflammation with neutrophils, transepithelial and transendothelial migration; and analysis of blood-brain barrier, dendritic cells, and Macrophages
  • Pathways for stem cell differentiation
  • Screening for population-specific neuronal motogens
  • Migration of normal, transformed and transfected cells
  • Chemoinvasion assays, drug discovery
Description & Specifications

Specifications

Yes
0.08cm2
Insert plate with 96 square well, flat bottom plate and lid
PET
8μm
Yes
Yes
Fluorescence plate reader or inverted fluorescence microscope
Polystyrene
96
1/Cs.
FluoroBlok™ HTS 96 Well Multiwell Permeable Support System