Corning™ Falcon™ Optilux 384-Well Microtest Microplates
Manufactured from polystyrene and treated with tissue culture to optimize cell growth. Corning™ Falcon™ Optilux 384-Well Microtest Microplates are ideal for use in high throughput systems.
$1,082.46 - $1,407.87
Specifications
Lid Yes
Material Polystyrene
Sterile Yes
Type 384-Well Plate
Volume (Metric) Well 130μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
08-772-116 Corning™
C353963
White with clear bottom Case of 50 Each for $1,406.64
08-772-115 Corning™
C353962
Black with clear bottom Case of 50 Each for $1,407.87
08-772-129 Corning™
C353988
Solid white Case of 50 Each for $1,190.00
08-772-114 Corning™
C353961
Clear Case of 50 Each for $1,082.46
Specifications
Description
  • Tissue culture (TC) treated for cell attachment
  • Beveled-edge square wells
  • Meet standard ANSI/SBS footprint dimensions for microplates
  • Indefinite shelf life
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Description & Specifications
Specifications
Yes
Yes
130μL
Polystyrene
384-Well Plate
15 to 110μL