missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Falcon™ FluoroBlok™ 24 Well Permeable Supports and Companion Plates

Block light transmission, simplifying detection of migrating fluorescently labeled cells. Corning™ Falcon™ FlouroBlok™ 24 Well Plate Permeable Supports are designed with a unique light-tight PET membrane. Must be used with Falcon™ 24 Well Companion Plate.

$537.17 - $1,631.01

Specifications

Sterility Sterile
Well Count 24
Material Polystyrene
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Type Pore Size Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Type Pore Size Price Quantity  
08-772-141 View Documents Corning™
C351151
Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate FluoroBlok™ Cell Culture Inserts for 24-Well plates 3 μm Case of 48 Each for $1,631.01
 
08-772-142 View Documents Corning™
C351152
Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate FluoroBlok™ Cell Culture Inserts for 24-Well plates 8μm Case of 48 Each for $1,631.01
 
08-771-25 View Documents Corning™
C353504
Falcon™ or Corning™ BioCoat™ Cell culture Inserts; may be used with or without cell culture inserts Falcon™ 24-Well TC-Treated Cell Polystyrene Permeable Support Companion Plate Case of 50 Each for $537.17
 
Description

Description

  • Dyed PET microporous membrane efficiently blocks the transmission of light within the range of 400-700 nm
  • Fluorescently labeled cells present in the top chamber of the insert are made invisible by the Corning™ FluoroBlok™ membrane
  • Migrated cells are no longer shielded from the light source and are easily detected with a fluorescence plate reader or inverted fluorescence microscope
  • Store at room temperature, away from direct sunlight.
Applications
  • Study a wide range of cell types and activities such as:
  • Inflammation with neutrophils, transepithelial and transendothelial migration; and analysis of blood-brain barrier, dendritic cells, and Macrophages
  • Pathways for stem cell differentiation
  • Screening for population-specific neuronal motogens
  • Migration of normal, transformed and transfected cells
  • Chemoinvasion assays, drug discovery
NOTE: Individual inserts must be used with Falcon Cell Culture Insert Companion plates to properly position the inserts in the wells. These plates accurately fit the Corning FluoroBlok 24 Multiwell insert. The Corning FluoroBlok 96 Multiwell inserts use the supplied square well receiver plates. These plates must be used for running the assay, labeling and reading samples to achieve reliable assay results.
Specifications

Specifications

Sterile
24
Polystyrene