Please login to your online account to display your discounted pricing

Corning™ Falcon™ FluoroBlok™ 24 Well Permeable Supports and Companion Plates


Block light transmission, simplifying detection of migrating fluorescently labeled cells. Corning™ Falcon™ FlouroBlok™ 24 Well Plate Permeable Supports are designed with a unique light-tight PET membrane. Must be used with Falcon™ 24 Well Companion Plate.

Manufacturer: corning™  C351151

 View more versions of this product

Catalog No. 08-772-141

  • / Case of 48 Each
 

Description & Specifications

Specifications

Closure Yes
For Use With (Equipment) Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate
Type FluoroBlok™ Cell Culture Inserts for 24 well plates
Culture Area 0.3cm2
Diameter (English) Membrane 0.25 in.
Diameter (Metric) Membrane 0.65cm
Format 24-well
Material Polystyrene
Membrane Polyethylene Terephthalate
No. of Wells 24
Pore Size 3μm
Sterile Yes
Format 24-well
Wells 24
Material Polystyrene
Type FluoroBlok Cell Culture Inserts
Lid No

  • Dyed PET microporous membrane efficiently blocks the transmission of light within the range of 400-700 nm
  • Fluorescently labeled cells present in the top chamber of the insert are made invisible by the Corning™ FluoroBlok™ membrane
  • Migrated cells are no longer shielded from the light source and are easily detected with a fluorescence plate reader or inverted fluorescence microscope
  • Store at room temperature, away from direct sunlight.
Applications
  • Study a wide range of cell types and activities such as: 
  • Inflammation with neutrophils, transepithelial and transendothelial migration; and analysis of blood-brain barrier, dendritic cells, and Macrophages
  • Pathways for stem cell differentiation
  • Screening for population-specific neuronal motogens
  • Migration of normal, transformed and transfected cells
  • Chemoinvasion assays, drug discovery
NOTE: Individual inserts must be used with Falcon Cell Culture Insert Companion plates to properly position the inserts in the wells.  These plates accurately fit the Corning FluoroBlok 24 Multiwell insert. The Corning FluoroBlok 96 Multiwell inserts use the supplied square well receiver plates. These plates must be used for running the assay, labeling and reading samples to achieve reliable assay results.