Please login to your online account to display your discounted pricing

FAK Inhibitor 14, Tocris™

Selective FAK inhibitor

$139.08 - $574.46

Chemical Identifiers

Chemical Name or Material FAK Inhibitor 14
Assay Percent Range >98%
Molecular Formula C6H10N4·4HCl
Formula Weight 284.01
Synonym 1,2,4,5-Benzenetetramine tetrahydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

341450

 
r & d systems
RDS341450
50mg Each for $574.46

341410

 
r & d systems
RDS341410
10mg Each for $139.08
Description & Specifications

Specifications

Formula Weight 284.01
Quantity 50mg
Chemical Name or Material FAK Inhibitor 14
Assay Percent Range >98%
CAS 4506-66-5
Synonym 1,2,4,5-Benzenetetramine tetrahydrochloride
Molecular Formula C6H10N4·4HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature