missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

F4/80, clone: BM8, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS14480182

 View more versions of this product

Catalog No. 501129661



Description

Description

EGF-Like Module Containing Mucin-Like Hormone Receptor-Like Sequence 1 (EMR1) is characterized as an Orphan-B GPCR with an unknown ligand. The C-terminus contains seven hydrophobic segments, characteristic of a G protein-coupled receptor. The N-terminus of EMR1 has 6 EGF-like modules, separated from the transmembrane segments by a serine/threonine-rich domain, a feature reminiscent of mucin-like, single-span, integral membrane glycoproteins with adhesive properties. EMR1 has been reported to be expressed in human peripheral blood mononuclear cells, monocytes, and hematopoietic and solid tumor-derived cell lines. ESTs have been isolated from human normal spleen and mixed liver/spleen libraries and from human kidney, colon, brain, and bone marrow cancer libraries.

This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The BM8 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded mouse tissue using pronase treatment and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
Specifications

Specifications

F4/80
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q61549
DD7A5-7, EGF-TM7, F4/80, Gpf480, Ly71, TM7LN3
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
BM8
Unconjugated
Adgre1
Liquid
Adgre1
IgG2a, kappa
100 μg
4° C
Primary
13733
Documents

For Research Use Only.