missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
F3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002152B01P
Description
Sequence: METPAWPRVPRPETAVARTLLLGWVFAQVAGASGTTNTVAAYNLTWKSTNFKTILEWEPKPVNQVYTVQISTKSGDWKSKCFYTTDTECDLTDEIVKDVKQTYLARVFSYPAGNVESTGSAGEPLYENSPEFTPYLETNLGQPTIQSFEQVGTKVNVTVEDERTLVRRNNTFLSLRDVFGKDLIYTLYYWKSSSSGKKTAKTNTNEFLIDVDKGENYCFSVQAVIPSRTVNRKSTDSPVECMGQEKGEFREIFYIIGAVVFVVIILVIILAISLHKCRKAGVGQSWKENSPLNVSSpecifications
F3 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_001993 | |
F3 | |
F3 (AAH11029.1, 1 a.a. ∼ 295 a.a) full-length human protein. | |
RUO | |
2152 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Flow Cytometry,Western Blot | |
Unconjugated | |
coagulation factor III (thromboplastin, tissue factor) | |
CD142/TF/TFA | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |