Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody


Specifically detect ezrin in human, monkey, cow, kangaroo, rat, mouse and hamster samples with Thermo Scientific™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS661P1ABX

 View more versions of this product

Catalog No. MS661P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Ezrin/p81/80K/Cytovillin Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone 3C12
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Kangaroo
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen A recombinant fragment of human ezrin corresponding to aa 362-585
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Bovine
Target Species Hamster
Target Species Human
Target Species Kangaroo
Target Species Murine
Target Species Non-human Primate
Target Species Rat

Specifically detect ezrin in human, monkey, cow, kangaroo, rat, mouse and hamster samples with Thermo Scientific™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody.

Ezrin (also known as p81, 80K, or cytovillin) is a cytoplasmic protein enriched in microvilli and other cell surface structures. Its NH2-terminal domain is associated with the plasma membrane and the COOH-terminal domain with the cytoskeleton. Ezrin has an actin-binding capability. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas. Ezrin serves as a tyrosine kinase substrate, and is phosphorylated in EGF-stimulated cells.

Host Species: Mouse

Clone: 3C12

Isotype: IgG1

Species Reactivity: Human, Monkey, Cow, Kangaroo, Rat, Mouse and Hamster. Others not known.

Epitope: aa 362-585

Immunogen: A recombinant fragment of human ezrin corresponding to aa 362-585

Molecular Weight: 81 kDa

Positive Control: Raji cells, Lung

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO