missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ eZKine™ 4-Color Compensation Control Kit

Designed for compensation setup with eZKine Whole Blood Intracellular Cytokine Kits. Contains necessary components to create single-color compensation controls for use with eZKine cocktails, including OneComp eBeads and 4 vials of antibody representing the 4 fluorochromes used in eZKine cocktails.

Supplier:  Invitrogen™ 8822685641

Catalog No. 501125103


Alert:

Contains Sodium azide


Includes: OneComp eBeads: 1 drop/test, 100 tests
Anti-Human CD3 (SK7) APC: 25 tests
Anti-Human CD4 (SK3) PerCP-eFluor 710: 25 tests
Anti-Human IL-17A (eBio64DEC17) PE: 25 tests
Anti-Human IFN gamma (4S.B3) FITC: 25 tests

Description

Description

OneComp eBeads can be stained with each of the 4 fluorochrome-conjuated antibodies to set up single-color compensation control tubes.
Specifications

Specifications

Human
CD3, CD4, IFN gamma, IL-17A
Flow Cytometric Analysis
2°C to 8°C. Do not freeze. Light sensitive material.
SDS
Documents

Documents

Promotions

Promotions