Please login to your online account to display your discounted pricing

Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin and Labeling Kits


Biotinylate protein amines (e.g., lysines) with our shortest (13.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Manufacturer: thermo scientific™  21326

 View more versions of this product

Catalog No. PI21326

  • / Each
 

Description & Specifications

Specifications

Quantity 8 x 1mg
Cleavable No
PEGylated No
Spacer Arm Length 13.5
Description EZ-Link Sulfo-NHS-Biotin, No-Weigh Format
Water Soluble Yes
Storage Requirements -20°C with desiccant
Chemical Reactivity Amine
Formulation Sulfo-NHS ester of biotin
Molecular Weight 443.43
Membrane Permeable No
Sufficient For 8 uses to label a total of 120mg of IgG using typical conditions

Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.

Highlights:

  • Shorter spacer arm than Sulfo-NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate, or phosphate buffers
  • Negative charge of -SO3 keeps biotinylation localized at the cell surface so reagent does not pass through cell membrane
  • Water solubility eliminates the need for harmful organic solvents
U.S. Patents 5,892,628 and 5,872,261.