Please login to your online account to display your discounted pricing

Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin and Labeling Kits


Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Manufacturer: thermo scientific™  PI21329

 View more versions of this product

Catalog No. PI21329

  • / Each
 

Description & Specifications

Specifications

Formulation NHS ester of biotin with a tetraethyleneglycol spacer
Spacer Arm Length 29
Cleavable No
Membrane Permeable No
Quantity 8 x 2mg
Chemical Reactivity Amine
PEGylated Yes
Molecular Weight 588.67
Water Soluble Yes
Storage Requirements Store in original bottle at 4°C with desiccant
Sufficient For 8 uses to label a total of 181mg of IgG using typical conditions
Description EZ-Link NHS-PEG4-Biotin, No-Weigh Format
Item Description No-Weigh; 8 x 2mg

Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Highlights:

  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Prevents aggregation of antibodies better than Sulfo-NHS-LC-Biotin
  • Labels at N-terminus or lysine residues in less than 1 hour
  • Thermo Scientific No-Weigh Packaging (8 x 2mg) saves time - preserves reagent activity and prevents cross-contamination between use