Please login to your online account to display your discounted pricing

Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin


Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.

Manufacturer: thermo scientific™  PI21902BID

 View more versions of this product

Catalog No. PI21902

  • / Each
 

Description & Specifications

Specifications

Membrane Permeable No
PEGylated Yes
Spacer Arm Length 29.1
Quantity 8 x 2mg
Water Soluble Yes
Chemical Reactivity Sulfhydryl
Cleavable No
Description EZ-Link Maleimide-PEG2-Biotin, No-Weigh Format
Formulation Maleimide-activated biotin with diethyleneglycol spacer
Molecular Weight 525.62
Sufficient For 8 uses to label a total of 203mg of IgG using typical conditions

Thermo Scientific™ EZ-Link Maleimide-PEG2-Biotin is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.

Highlights:

  • Water-soluble; contains polyethylene glycol (PEG) spacer arm
  • Reacts with sulfhydryl (-SH) groups at acidic to neutral pH
  • Transfers hydrophilic PEG spacer arm to final conjugate
  • Forms a thioether bond