missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Extran™ MA 01

Use for manual cleaning. MilliporeSigma Extran™ MA 01 is an alkaline concentrated liquid. Extran cleaning agents have a consistent composition and are biodegradable, with no residues, impurities, harsh chemicals or scents. (Note: this product is iden

Manufacturer:  MilliporeSigma™ 1075555000

 View more versions of this product

Catalog No. M1075555000



Specifications

Specifications

5L
Colorless
12.94 (undiluted), 11–12 (5%, in water)
1.07 g/cm3 (20°C)
Manual cleaning
<100°C
Plastic
Concentrated liquid
<23.4 hPa (20°C)
Documents
Promotions

Promotions