missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Extran™ AP 41

Recommended for use on glass, stainless steel, porcelain, plastic, rubber, and fiberglass. MilliporeSigma Extran™ AP 41 is an enzymatic powdered cleaner designed to remove mucus, saliva, blood, and protein residues. Dissolve 3g/L and use at 60°C

$284.89 - $2,441.49

Specifications

pH Range 11.4 (3g/L in water, 20°C)
Format Powdered solid
For Use With (Application) Remove mucus, saliva, blood, and protein residues
Color White
Composition Enzymes, phosphates, and alkali salts
View More Specs

Products 2
Catalog Number Mfr. No. Size Container Type Price Quantity  
Catalog Number Mfr. No. Size Container Type Price Quantity  
M1075702000 View Documents MilliporeSigma™
1075702000
2kg Plastic bottle Each for $284.89
 
M1075709025 View Documents MilliporeSigma™
1075709025
25kg Plastic drum Each for $2,441.49
 
Specifications

Specifications

11.4 (3g/L in water, 20°C)
Remove mucus, saliva, blood, and protein residues
Enzymes, phosphates, and alkali salts
Powdered solid
White