MilliporeSigma™ Extran™ AP 22
Use when gentle conditions are required, e.g. removing deposits on taps, glass and sensitive metals. MilliporeSigma Extran™ AP 22 is an acidic concentrated liquid with citric acid. It works as a pre-wash, as a main wash to remove amines and proteins,
Manufacturer:  MilliporeSigma™ 1075619025
 View more versions of this product
Catalog No. M1075619025
Specifications
Specifications
25L
Citric acid
Plastic container
Concentrated liquid
Prewash, Cleaning in automatic washers, Neutralizing rinse after alkaline washing
Colorless
32.0–35.0%
3.0 (2g/L in water, 20°C)
1.15 g/cm3 (20°C)
Documents