MilliporeSigma™ Extran™ AP 12
Remove all types of contamination, especially starch and protein residues. MilliporeSigma Extran™ AP 12 is an alkaline powdered cleaning agent for automatic washers. Extran products are biodegradable, with no residues, impurities or harsh chemicals.
Manufacturer:  MilliporeSigma™ 1075639010
 View more versions of this product
Catalog No. M1075639010
Specifications
Specifications
10kg
Plastic bucket
Powdered solid
Automatic washing
White
12.3 (3g/L in water, 20°C)
900 kg/m3