missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Extraction Solvent, 4mL/L Hydrochloric Acid in Reagent Alcohol, LabChem™

$115.11 - $115.11

Chemical Identifiers

CAS 64-17-5,67-63-0,67-56-1,7647-01-0
CAS Max % 89.6,5,5,0.4
Linear Formula HCl
Formula Weight 36.46
UN Number UN1993
Chemical Name or Material Extraction Solvent, 4mL/L Hydrochloric Acid in Reagent Alcohol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC141804 Lab Chem
LC141804
4L Each for $115.11
Specifications Description & Specifications

Chemical Identifiers

64-17-5,67-63-0,67-56-1,7647-01-0
HCl
UN1993
89.6,5,5,0.4
36.46
Extraction Solvent, 4mL/L Hydrochloric Acid in Reagent Alcohol

Specifications

64-17-5,67-63-0,67-56-1,7647-01-0
Liquid
36.46
Passes Test
Soluble in water
Hydrogen chloride; Carbon monoxide; Carbon dioxide; May release flammable gases
4L
89.6,5,5,0.4
HCl
UN1993
Poly Bottle
Room Temperature
Clear, colorless
Extraction Solvent, 4mL/L Hydrochloric Acid in Reagent Alcohol

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
May cause cancer.
Suspected of damaging fertility or the unborn child (oral).
Causes damage to organs (liver, kidneys, central nervous system, optic nerve).

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger