Please login to your online account to display your discounted pricing
Thermo Scientific™ External-Controlled Hotplates (Largest)
Enable uniform heating of large volumes and multiple vessels using External-Controlled Hotplates, ideal for a range of applications with acidic or basic aqueous solutions.
$2,351.75 - $2,960.24
Specifications
Amperage 13.3A
Hertz 50/60Hz
Temperature Range (Metric) 150° to 371°C
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Voltage Price Quantity    
1149666Q
  Accessories
thermo scientific™
RC2235Q
120V Each for $2,351.75
114966EQ
  Accessories
thermo scientific™
RC2240Q
240V Each for $2,960.24
Description & Specifications
Specifications
Amperage 13.3A
Hertz 50/60Hz
Temperature Range (Metric) 150° to 371°C
Enable uniform heating of large volumes and multiple vessels using Thermo Scientific™ External-Controlled Hotplates. These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environment.
Key Features:
  • Isolation of controls allows operation in hazardous environments
  • Large 12 x 12″ (30.5 x 30.5cm) and 12 x 24″ (30.5 x 60.9cm) top plate surfaces accommodate up to 40 lb. (18.1kg) loads
  • Power light indicates when control is operating
Temperature Control:
  • Large capacity with excellent temperature stability
  • Temperature stability is ±5°C (9°F) over the operating temperature range Embedded heating elements maximize temperature uniformity and evenly transfer heat within ±10°C (18°F) at 150°C (300°F)
  • Percentage input controller delivers infinite temperature selection
Construction:
  • Porcelain-coated, stainless-steel top offers chemical and stain resistance
  • Chemical-resistant cover protects line cord
  • Stainless-steel case offers maximum resistance to corrosion
Includes:
5′ (1.5m) 3-wire cord and plug