missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ External-Controlled Hot Plate, 150°C to 371°C, Porcelain-coated Stainless Steel

Enable uniform heating of large volumes and multiple vessels.

Supplier:  Thermo Scientific™ RC2235Q

 View more versions of this product

Catalog No. 1149666Q



Description

Description

Enable uniform heating of large volumes and multiple vessels using Thermo Scientific™ External-Controlled Hotplates. These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environment.

Key Features:
  • Isolation of controls allows operation in hazardous environments
  • Large 12 x 12″ (30.5 x 30.5 cm) top plate surface accommodates up to 40 lb. (18.1 kg) loads
  • Power light indicates when control is operating
Temperature Control:

  • Large capacity with excellent temperature stabilityy
  • Temperature stability is ±5&°C;C (9&°C;F) over the operating temperature range Embedded heating elements maximize temperature uniformity and evenly transfer heat within ±10&°C;C (18&°C;F) at 150&°C;C (300&°C;F)
  • Percentage input controller delivers infinite temperature selection

Construction:
  • Porcelain-coated, stainless-steel top offers chemical and stain resistance
  • Chemical-resistant cover protects line cord
  • Stainless-steel case offers maximum resistance to corrosion

Includes:

5′ (1.5 m) 3-wire cord and plug

Specifications

Specifications

12 in. x 24 in.
Hot Plate
120 V
Porcelain-coated Stainless Steel
150°C to 371°C
13.3 A
50/60 Hz
18 kg
COA, COC
Rectangle
12 x 12 in.
1600 w
120 V, 50/60 Hz
SDS
Documents

Documents

Accessories
Promotions

Promotions