Please login to your online account to display your discounted pricing
Thermo Scientific Expression Vectors for Pierce™ qIP Kits
Optimize cloning with this group of vectors, which includes epitope-tagged (HA and c-Myc) and TurboLuc-tagged (Tluc) multiple cloning site mammalian expression vectors.
Manufacturer: thermo scientific  82017
 View more versions of this product
Catalog No. PI82017
  • / Each
Description & Specifications
Specifications
Quantity 10μg
Formulation 0.5μg/μL DNA in 10mM Tris-HCl, pH 8.5
Quantity 10μg
Optimize cloning in the Thermo Scientific ™ Pierce™ quantitative IP (qIP) protein interaction system with this group of Thermo Scientific™ pCMV Vectors, which includes epitope-tagged (HA and c-Myc) and TurboLuc-tagged (Tluc) multiple cloning site mammalian expression vectors.