Please login to your online account to display your discounted pricing

Thermo Scientific Explosion-Proof Refrigerator/Freezers


Prevent the triggering of an explosion inside or outside of the unit with Thermo Scientific™ Explosion-Proof Refrigerator/Freezers.

Manufacturer: thermo scientific  10ECEETSA

 View more versions of this product

Catalog No. 10ECEETSA

  • / Each
 

Description & Specifications

Specifications

Dimensions (L x W x H) Exterior 24 x 23.63 x 59.75 in. (60.96 x 60 x 151.77cm)
Temperature Range (Metric) Ref: 1°C to 12°C Frz: -12°C to -20°C
Voltage 115V
Certifications/Compliance UL listed meets NFPA 45 and 70
Height (Metric) Exterior 152.4cm
Electrical Requirements 115 V/60 Hz
Height (Metric) Refrigerator 99cm
Control Mechanical Dial Thermostat/Outside on Rear of Unit
Wattage 83w
Warranty 13 Months Parts and Labor
For Use With (Application) Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
Typical Lifetime 10 Years
Min. Clearance 2 in. On Sides, 2 in. At Back, 2 in. On Top
Lighting Interior - none
Temperature (Setpoint) Adjustable
Current 1.9Amps/230Watts
Shelves 3 Adjustable Interior, 1 Bottom Drawer, 4 Door Shelves
Amperage 1.2A
Shipping Weight (English) 143 lb.
Shipping Weight (Metric) 65kg
Temperature Range (Freezer) -12° to -20°C
Temperature Range (Refrigerator) 1° to 12°C
Width (English) Exterior 23.5 in.
Width (English) Freezer 18 in.
Width (Metric) Refrigerator 49.5cm
Defrost Manual
Amperage 1.2A
Capacity (English) 10 cu. ft.
Capacity (Metric) 283.17L
Compressor Bottom Mounted
Depth (English) Freezer 16.25 in.
Depth (Metric) Exterior 77.5cm
Display Temperature - none
Depth (English) Refrigerator 18 in.
Exterior Finish Enameled steel
Insulation Polyurethane Foam
Depth (Metric) Freezer 41.3cm
Capacity 10 cu. ft. / 283.17L
Dimensions (D x W x H) Interior Rfgr: 18 x 19.5 x 39” (45.7 x 49.5 x 99cm); Frzr: 16.25 x 18 x 12” (41.3 x 45.7 x 30.5cm)
Hertz 60Hz
Depth (Metric) Refrigerator 45.7cm
Interior Finish ABS Plastic
Insulation Polyurethane Foam
Model 10ECEETSA
Net Weight (Metric) 54.5kg
Plug Type None supplied
Refrigerant R134a, 100% CFC Free
Height (English) Freezer 12 in.
Type Explosion Proof Freezer/Refrigerator
Height (English) Refrigerator 39 in.
Color White
Height (Metric) Freezer 30.5cm
Capacity 10.1 Cubic Feet/286 Liters
Width (English) Refrigerator 19.5 in.
Width (Metric) Exterior 59.7cm
Width (Metric) Freezer 45.7cm
Capacity (English) 10.1 cu. ft.
Capacity (Metric) 286L
Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Chamber Material Enameled steel, White
Depth (English) Exterior 30.5 in.
Doors 2 (2 compartments); Hinged, Open Left to Right, Reversible
Door Count 2(2 compartments)
Door Style Hinged, Open Left to Right, Reversible
Net Weight (English) 120 lb.
Height (English) Exterior 60 in.

Prevent the triggering of an explosion inside or outside of the unit with Thermo Scientific™ Explosion-Proof Refrigerator/Freezers. Suitable for use in Class I, Division I Group C and D hazardous environments, this device meets OSHA and National Fire Protection Association safety requirements.

  • Thermostat and compressor components are isolated within a vapor-proof enclosure
  • Electrical connections are hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
  • Manual defrost

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.