Please login to your online account to display your discounted pricing

Thermo Scientific Explosion-Proof Refrigerator/Freezers

Prevent the triggering of an explosion inside or outside of the unit with Thermo Scientific™ Explosion-Proof Refrigerator/Freezers.

$8,555.71 - $8,555.71

Specifications

Capacity 10.1 Cubic Feet/286 Liters
Certifications/Compliance UL listed meets NFPA 45 and 70
Defrost Ref: Automatic Defrost; Frz: Manual Defrost
Refrigerant R134a, 100% CFC Free
Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear
View More Specs
Products
Catalog Number Mfr. No. Electrical Requirements Capacity Price Quantity    

10ECEETSA

 
thermo scientific
10ECEETSA
115 V/60 Hz 10.1 Cubic Feet/286 Liters Each for $8,555.71
Description & Specifications

Specifications

Capacity 10.1 Cubic Feet/286 Liters
Certifications/Compliance UL listed meets NFPA 45 and 70
Defrost Ref: Automatic Defrost; Frz: Manual Defrost
Refrigerant R134a, 100% CFC Free
Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Compressor Bottom Mounted
Door Count 2(2 compartments)
Door Style Hinged, Open Left to Right, Reversible
Temperature Range (Refrigerator) 1° to 12°C
Temperature Range (Freezer) -12° to -20°C
Exterior Finish Enameled steel
Dimensions (L x W x H) Exterior 24 x 23.63 x 59.75 in. (60.96 x 60 x 151.77cm)
Interior Finish ABS Plastic
Insulation Polyurethane Foam
Shelves 3 Adjustable Shelves, 1 Fixed Shelf, 1 Drawer, 4 Door Shelves

Prevent the triggering of an explosion inside or outside of the unit with Thermo Scientific™ Explosion-Proof Refrigerator/Freezers. Suitable for use in Class I, Division I Group C and D hazardous environments, this device meets OSHA and National Fire Protection Association safety requirements.

  • Thermostat and compressor components are isolated within a vapor-proof enclosure
  • Electrical connections are hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal CFC-free insulation and refrigerant
  • Manual defrost

13 Months Parts and Labor

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.