missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Explosion-Proof Refrigerator
Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.
Manufacturer: Thermo Scientific™ 05EREETSA
Description
Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.
- Hydraulic thermostat
- Manual defrost
- Magnetic vinyl gaskets for positive door seal
- CFC-free insulation and refrigerant
Specifications
Door Lock | |
White | |
Explosion-Proof Refrigerator | |
5.5 cu. ft. | |
Enameled Steel | |
17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm) | |
1° to 12°C | |
115V | |
115 V/60 Hz | |
100 lb. | |
160 Watts | |
Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments | |
4 in. On Sides, 3 in. At Back, 4 in. On Top | |
Adjustable | |
2 Adjustable Feet In Front, 2 Fixed Pads In Rear | |
Temperature Display - None | |
Polyurethane Foam | |
R134a, 100% CFC Free | |
120 lb. | |
Manual |
UL listed meets NFPA 45 and 70 | |
Mechanical Dial Thermostat/Outside on Rear of Unit | |
1.0 Amps/160 Watts | |
155.7L | |
23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1 cm) | |
Not Provided | |
Explosion-Proof Refrigerator | |
1(1 compartment) | |
ABS Plastic | |
45.3kg | |
13 Months Parts and Labor | |
10 Years | |
Interior - No | |
1.33A | |
Bottom Mounted | |
60Hz | |
05EREETSA | |
3 Adjustable Shelves, Storage On Bottom Of Unit | |
54.5kg | |
Hinged, Opens Left to Right, Reversible |