missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Explosion-Proof Freezers

Maintain a safe work environment even in hazardous conditions with this explosion-proof freezer.

Manufacturer:  Thermo Scientific™ 05EFEETSA

 View more versions of this product

Catalog No. 05EFEETSA



Description

Description

Thermo Scientific explosion-proof laboratory refrigerators, freezers and combinations units are particularly suited for safe storage in hazardous environments.

Features:
  • Temperature range: -20° to -12°C
  • Thermostat and compressor components isolated within a vapor-proof enclosure
  • Electrical connections hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
  • Hydraulic thermostat
  • Two fixed shelves, three door shelves
  • Aluminum evaporator coil
Specifications

Specifications

UL listed meets NFPA 45 and 70
Mechanical Dial Thermostat/Outside on Rear of Unit
5 cu. ft.
Enameled Steel
19 x 18.63 x 29.5 in. (48.2 x 47.32 x 74.9)
1° to 12°C
115V
115 V/60 Hz
Warranty varies by country, please contact us for details
Epoxy-enameled steel, White
Bottom Mounted
38.1cm Upper; 25.4cm Bottom
28 in.
45.7cm
Polyurethane Foam
75.9cm
R134a, 100% CFC Free
120 lb.
18 in.
Manual
21.5 in.
White
1.1Amps
141.6L
25.75 x 21.38 x 33.13 in. (84.15 x 54.3 x 65.4)
Not Provided
Explosion-Proof Freezer
1 (1 compartment)
ABS Plastic
1.6A
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
15 in. Upper; 10 in. Bottom
33.5 in.
85.1cm
60Hz
29.5 in.
05EFEETSA
2 Fixed Shelves, 1 Bottom Tray, 3 Door Shelves
54.49kg
45.7cm
Hinged, Opens Left to Right, Non-Reversible
54.6cm
Documents