missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Exploring Life Poster Set

Vibrant posters feature highly detailed illustrations and photographs.

Manufacturer:  Fisher Science Education AZ-35-1045

 View more versions of this product

Catalog No. S66677


 

Specifications Description & Specifications

Specifications

Science
35 in.
7 Informative posters like exploring with a microscope, visualizing cells, visualizing mitosis, living sands, Human genetic disorders, visualizing Blood, systems of the Human body
Exploring Life Poster Set
58.42cm
Highly detailed illustrations and photographs
88.9cm
Paper
23 in.