missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Exploring Life Poster Set

Vibrant posters feature highly detailed illustrations and photographs.

$99.25 - $148.50

Specifications

Classification Science
For Use With (Application) Highly detailed illustrations and photographs
Height (English) 35 in.
Height (Metric) 88.9cm
Includes 7 Informative posters like exploring with a microscope, visualizing cells, visualizing mitosis, living sands, Human genetic disorders, visualizing Blood, systems of the Human body
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
S66677 Fisher Science Education
AZ-35-1045
Paper Each for $99.25
S66678 Fisher Science Education
AZ-35-1046
Laminated Each for $148.50
Specifications

Description

  • Explores several life science topics
  • Dimensions: 23 × 35 in. (58.4 × 88.9cm)

Set includes seven informative posters:

  • Exploring with a microscope
  • Visualizing cells
  • Visualizing mitosis
  • Living sands
  • Human genetic disorders
  • Visualizing blood
  • Systems of the human body

Description & Specifications

Specifications

Science
35 in.
7 Informative posters like exploring with a microscope, visualizing cells, visualizing mitosis, living sands, Human genetic disorders, visualizing Blood, systems of the Human body
58.42cm
Highly detailed illustrations and photographs
88.9cm
23 in.