Please login to your online account to display your discounted pricing

Exploring Cell Processes Lab Investigation

A complete kit for plant and animal cell observation.

$113.60 - $186.30

Specifications

For Use With (Application) Identify and compare organelles in plant and animal cells, relating their structure to their specific function
Includes Stain set, microscope slides, coverslips, and mounting media, dialysis tubing and buffer, starch, sucrose, and starch indicator, test slips, comprehensive teacher and student guides
View More Specs
Includes: 
  • Stain set
  • Microscope slides, coverslips, and mounting media
  • Dialysis tubing and buffer
  • Starch, sucrose, and starch indicator
  • Test slips
  • Comprehensive teacher and student guides
Requires:  Compound microscopes
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S63070

 
fisher science education
AZ-20-1203
Exploring Cell Processes Kit Each for $113.60

S66699

 
fisher scientific
AZ-25-1111
Exploring Cell Processes Each for $186.30
Description & Specifications

Specifications

For Use With (Application) Identify and compare organelles in plant and animal cells, relating their structure to their specific function
Includes Stain set, microscope slides, coverslips, and mounting media, dialysis tubing and buffer, starch, sucrose, and starch indicator, test slips, comprehensive teacher and student guides

Students learn how to make observations and comparisons, analyze data, relate cause and effect, and make inferences.

  • Compare plant and animal cells
  • Illustrate cell structure and processes to include mitochondria, cell membranes, DNA and RNA identification, osmosis, plasmolysis, and more
  • Identify and compare organelles in plant and animal cells, relating their structure to their specific function

Class size: 40 students
Great value! Save money when you buy this kit with its corresponding CD-ROM (S66589) as a curriculum pack