Please login to your online account to display your discounted pricing

Exploring with a Microscope


Study a whole microworld.

Manufacturer: fisher science education  AZ-20-1063

 View more versions of this product

Compound microscopes

Catalog No. S63072

  • / Each
 

Description & Specifications

Specifications

Includes Magnifiers, nut and bolt microtomes, paraffin, pipets, polarizing filters, pond mix, rulers, slides, stains, teacher and student guides
Product Type Microscope Kit
For Use With (Application) Study a whole microworld
Kit Size 40 students
Class Size 40 students